Assay Details
Gene Family ID: | MIPF0000191, mir-340 |
Mature miRNA Sequence: | UCCGUCUCAGUUACUUUAUAGCC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Rat | rno-miR-340-3p | MIMAT0000585 | |
| Bovine | bta-miR-340 | MIMAT0009296 | |
| Cricetulus griseus | cgr-miR-340-3p | MIMAT0023921 | |
| Dasypus novemcinctus | dno-miR-340-3p | MIMAT0047824 | |
| Guinea pig | cpo-miR-340-3p | MIMAT0047179 | |
| Horse | eca-miR-340-3p | MIMAT0013067 | |
| View More Rows | |||
| Pteropus alecto | pal-miR-340-3p | MIMAT0040192 | |
| Rabbit | ocu-miR-340-3p | MIMAT0048387 | |
| Rhesus monkey | mml-miR-340-3p | MIMAT0026854 | |
Stem-loop Details
Rat
Bovine
Cricetulus griseus
Dasypus novemcinctus
Guinea pig
Horse
Pteropus alecto
Rabbit
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-340 (v9.2)
Mouse: mmu-miR-340 (v9.2)
Rat: rno-miR-340 (v9.2)
Mouse: mmu-miR-340 (v9.2)
Rat: rno-miR-340 (v9.2)

Genomic Map