Assay Details
Gene Family ID: | MIPF0000128, mir-450 |
Mature miRNA Sequence: | UUUUUGCGAUGUGUUCCUAAUA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Dog | cfa-miR-450a | MIMAT0001548 |
Stem-loop Details
Dog
More Information
Related Products
miRBase Alias:
Human: hsa-miR-450 (v9.2)
Mouse: mmu-miR-450 (v9.2)
Citrobacter farmeri: cfa-miR-450 (v10)
Mouse: mmu-miR-450 (v9.2)
Citrobacter farmeri: cfa-miR-450 (v10)

Genomic Map