Assay Details
Gene Family ID: | MIPF0000287, mir-452 |
Mature miRNA Sequence: | UCAGUCUCAUCUGCAAAGAAG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-452-3p | MIMAT0035749 |
Stem-loop Details
Rat
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | hsa-miR-452* |
|---|---|
| miRBase Accession Number: | MI0030363 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCAGUCUCAUCUGCAAAGAAG |
| Species: | Rat |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000287, mir-452 |
Mature miRNA Sequence: | UCAGUCUCAUCUGCAAAGAAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-452-3p | MIMAT0035749 |