More Information
Updated Mapping Information
miRBase Alias:
Mouse: mmu-miR-201 (v9.2)
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | mmu-miR-201 |
|---|---|
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UACUCAGUAAGGCAUUGUUCU |
| Product Type: | TaqMan™ MicroRNA Assay |