Assay Details
Mature miRNA Sequence: | UGUAGUGUUUCCUACUUUAUGG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Chicken | gga-miR-142-3p | MIMAT0001194 |
| Chrysemys picta | cpi-miR-142-3p | MIMAT0037767 |
| Gadus morhua | gmo-miR-142-2-3p | MIMAT0044046 |
| Ornithorhynchus anatinus | oan-miR-142-3p | MIMAT0006983 |
| Pig | ssc-miR-142-3p | MIMAT0020362 |
Stem-loop Details
Chicken
Chrysemys picta
Gadus morhua
Ornithorhynchus anatinus
Pig
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-142-3p (v9.2)
Ochrobactrum anthropi: oan-miR-142* (v18)
Ochrobactrum anthropi: oan-miR-142* (v18)

Genomic Map