Assay Details
Mature miRNA Sequence: | UUCCCUUUGUCAUCCUAUGCCUG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Python bivittatus | pbv-miR-204-5p | MIMAT0039006 |
Stem-loop Details
Python bivittatus
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | mmu-miR-204 |
|---|---|
| miRBase Accession Number: | MI0030278 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUCCCUUUGUCAUCCUAUGCCUG |
| Species: | Python bivittatus |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | UUCCCUUUGUCAUCCUAUGCCUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Python bivittatus | pbv-miR-204-5p | MIMAT0039006 |