Assay Details
Gene Family ID: | MIPF0000185, mir-542 |
Mature miRNA Sequence: | UCGGGGAUCAUCAUGUCACGAG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bovine | bta-miR-542-5p | MIMAT0013594 |
| Dasypus novemcinctus | dno-miR-542-5p | MIMAT0047969 |
| Pteropus alecto | pal-miR-542-5p | MIMAT0040183 |
Stem-loop Details
Bovine
Dasypus novemcinctus
Pteropus alecto

Genomic Map