Assay Details
Gene Family ID: | MIPF0000113, mir-188 |
Mature miRNA Sequence: | UACCCAUUGCAUAUCGGAGUUG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-660-5p | MIMAT0003338 | |
| Callithrix jacchus | cja-miR-660 | MIMAT0039432 | |
| Daubentonia madagascariensis | dma-miR-660 | MIMAT0049268 | |
| Dog | cfa-miR-660 | MIMAT0006760 | |
| Gorilla gorilla | ggo-miR-660 | MIMAT0024164 | |
| Horse | eca-miR-660 | MIMAT0013241 | |
| View More Rows | |||
| Pan paniscus | ppa-miR-660 | MIMAT0049103 | |
| Pan troglodytes | ptr-miR-660 | MIMAT0008308 | |
| Papio hamadryas | pha-miR-660 | MIMAT0049544 | |
| Pig | ssc-miR-660 | MIMAT0041612 | |
| Pongo pygmaeus | ppy-miR-660 | MIMAT0016092 | |
| Pteropus alecto | pal-miR-660-5p | MIMAT0039997 | |
| Rhesus monkey | mml-miR-660-5p | MIMAT0006499 | |
Stem-loop Details
Human
Callithrix jacchus
Daubentonia madagascariensis
Dog
Gorilla gorilla
Horse
Pan paniscus
Pan troglodytes
Papio hamadryas
Pig
Pongo pygmaeus
Pteropus alecto
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-660 (v17)
Macaca mulatta: mml-miR-660 (v19)
Macaca mulatta: mml-miR-660 (v19)

Genomic Map