Assay Details
Gene Family ID: | MIPF0000522, mir-601 |
Mature miRNA Sequence: | UGGUCUAGGAUUGUUGGAGGAG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-601 | MIMAT0003269 |
| Pan troglodytes | ptr-miR-601 | MIMAT0008270 |
| Pongo pygmaeus | ppy-miR-601 | MIMAT0016043 |
| Rhesus monkey | mml-miR-601 | MIMAT0006458 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus
Rhesus monkey

Genomic Map