Assay Details
Gene Family ID: | MIPF0000707, mir-764 |
Mature miRNA Sequence: | AGGAGGCCAUAGUGGCAACUGU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-764-3p | MIMAT0003895 |
| Horse | eca-miR-764-3p | MIMAT0013244 |
Stem-loop Details
Mouse
Horse

Genomic Map