Assay Details
Gene Family ID: | MIPF0000189, mir-345 |
Mature miRNA Sequence: | CCCUGAACUAGGGGUCUGGAGA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-345-3p | MIMAT0004655 |
| Pteropus alecto | pal-miR-345-3p | MIMAT0039982 |
Stem-loop Details
Rat
Pteropus alecto

Genomic Map