Assay Details
Gene Family ID: | MIPF0000220, mir-486 |
Mature miRNA Sequence: | CGGGGCAGCUCAGUACAGGAU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-486-3p | MIMAT0004762 |
| Mouse | mmu-miR-486a-3p | MIMAT0017206 |
| Horse | eca-miR-486-3p | MIMAT0013187 |
| Pteropus alecto | pal-miR-486a-3p | MIMAT0039847 |
| Rhesus monkey | mml-miR-486-3p | MIMAT0006345 |
| Tupaia chinensis | tch-miR-486-3p | MIMAT0036553 |
Stem-loop Details
Human
Mouse
Horse
Pteropus alecto
Rhesus monkey
Tupaia chinensis
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-486-3p (v20)
Mouse: mmu-miR-486* (v17)
Mouse: mmu-miR-486* (v17)

Genomic Map