Assay Details
Gene Family ID: | MIPF0000065, mir-132 |
Mature miRNA Sequence: | ACCGUGGCUUUCGAUUGUUACU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-132-5p | MIMAT0004594 | |
| Rat | rno-miR-132-5p | MIMAT0017123 | |
| Cricetulus griseus | cgr-miR-132-5p | MIMAT0023758 | |
| Dasypus novemcinctus | dno-miR-132-5p | MIMAT0047631 | |
| Guinea pig | cpo-miR-132-5p | MIMAT0046984 | |
| Monodelphis domestica | mdo-miR-132-5p | MIMAT0026693 | |
| View More Rows | |||
| Pteropus alecto | pal-miR-132-5p | MIMAT0039991 | |
| Rhesus monkey | mml-miR-132-5p | MIMAT0026816 | |
Stem-loop Details
Human
Rat
Cricetulus griseus
Dasypus novemcinctus
Guinea pig
Monodelphis domestica
Pteropus alecto
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-132* (v17)
Rat: rno-miR-132* (v18)
Rat: rno-miR-132* (v18)

Genomic Map