Assay Details
Gene Family ID: | MIPF0000190, mir-342 |
Mature miRNA Sequence: | AGGGGUGCUAUCUGUGAUUGA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-342-5p | MIMAT0004694 |
| Pongo pygmaeus | ppy-miR-342-5p | MIMAT0015844 |
| Pteropus alecto | pal-miR-342-5p | MIMAT0040085 |
| Rhesus monkey | mml-miR-342-5p | MIMAT0006282 |
Stem-loop Details
Human
Pongo pygmaeus
Pteropus alecto
Rhesus monkey

Genomic Map