Assay Details
Gene Family ID: | MIPF0000128, mir-450 |
Mature miRNA Sequence: | UUUUGCAAUAUGUUCCUGAAUA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-450b-5p | MIMAT0004909 | |
| Bovine | bta-miR-450b | MIMAT0024576 | |
| Gorilla gorilla | ggo-miR-450b | MIMAT0024296 | |
| Horse | eca-miR-450b-5p | MIMAT0013220 | |
| Pig | ssc-miR-450b-5p | MIMAT0013927 | |
| Pongo pygmaeus | ppy-miR-450b-5p | MIMAT0015894 | |
| View More Rows | |||
| Rabbit | ocu-miR-450b-5p | MIMAT0048460 | |
| Rhesus monkey | mml-miR-450b-5p | MIMAT0006333 | |
Stem-loop Details
Human
Bovine
Gorilla gorilla
Horse
Pig
Pongo pygmaeus
Rabbit
Rhesus monkey
More Information
Related Products
miRBase Alias:
Staphylococcus schleiferi: ssc-miR-450b (v15)

Genomic Map