Assay Details
Gene Family ID: | MIPF0000113, mir-188 |
Mature miRNA Sequence: | CCUCCCACACCCAAGGCUUGCA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-532-3p | MIMAT0004780 | |
| Mouse | mmu-miR-532-3p | MIMAT0004781 | |
| Rat | rno-miR-532-3p | MIMAT0005323 | |
| Dasypus novemcinctus | dno-miR-532-3p | MIMAT0047964 | |
| Guinea pig | cpo-miR-532-3p | MIMAT0047336 | |
| Horse | eca-miR-532-3p | MIMAT0013236 | |
| View More Rows | |||
| Pan troglodytes | ptr-miR-532 | MIMAT0008213 | |
| Pig | ssc-miR-532-3p | MIMAT0013941 | |
| Pongo pygmaeus | ppy-miR-532-3p | MIMAT0015988 | |
| Pteropus alecto | pal-miR-532-3p | MIMAT0039902 | |
| Rabbit | ocu-miR-532-3p | MIMAT0048520 | |
| Rhesus monkey | mml-miR-532-3p | MIMAT0006404 | |
Stem-loop Details
Human
Mouse
Rat
Dasypus novemcinctus
Guinea pig
Horse
Pan troglodytes
Pig
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Human Pool AMegaplex™ RT Primers, Rodent Pool A
Related Products

Genomic Map