Assay Details
Gene Family ID: | MIPF0000121, mir-202 |
Mature miRNA Sequence: | UUCCUAUGCAUAUACUUCUUUG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-202-5p | MIMAT0002810 |
| Chrysemys picta | cpi-miR-202-5p | MIMAT0037824 |
| Dog | cfa-miR-202 | MIMAT0009844 |
| Ophiophagus hannah | oha-miR-202-5p | MIMAT0036808 |
| Python bivittatus | pbv-miR-202-5p | MIMAT0039003 |
Stem-loop Details
Human
Chrysemys picta
Dog
Ophiophagus hannah
Python bivittatus

Genomic Map