Assay Details
Gene Family ID: | MIPF0000121, mir-202 |
Mature miRNA Sequence: | AGAGGUAUAGGGCAUGGGAA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-202-3p | MIMAT0002811 |
| Pan troglodytes | ptr-miR-202 | MIMAT0008064 |
| Taeniopygia guttata | tgu-miR-202 | MIMAT0014565 |
Stem-loop Details
Human
Pan troglodytes
Taeniopygia guttata

Genomic Map