Assay Details
Gene Family ID: | MIPF0000065, mir-132 |
Mature miRNA Sequence: | UAACAGUCUCCAGUCACGGCCA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-212-3p | MIMAT0000659 |
| Rat | rno-miR-212-3p | MIMAT0000883 |
| Guinea pig | cpo-miR-212-3p | MIMAT0047114 |
| Pteropus alecto | pal-miR-212-3p | MIMAT0039980 |
| Rabbit | ocu-miR-212-3p | MIMAT0048336 |
Stem-loop Details
Mouse
Rat
Guinea pig
Pteropus alecto
Rabbit
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-212 (v15)
Rat: rno-miR-212 (v18)
Rat: rno-miR-212 (v18)

Genomic Map