Assay Details
Gene Family ID: | MIPF0000545, mir-765 |
Mature miRNA Sequence: | UGGAGGAGAAGGAAGGUGAUG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-765 | MIMAT0003945 |
| Pan troglodytes | ptr-miR-765 | MIMAT0008320 |
| Pongo pygmaeus | ppy-miR-765 | MIMAT0016105 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus

Genomic Map