Assay Details
Gene Family ID: | MIPF0000667, mir-1249 |
Mature miRNA Sequence: | ACGCCCUUCCCCCCCUUCUUCA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-1249-3p | MIMAT0005901 | |
| Mouse | mmu-miR-1249-3p | MIMAT0010560 | |
| Rat | rno-miR-1249 | MIMAT0017892 | |
| Bovine | bta-miR-1249 | MIMAT0009976 | |
| Cricetulus griseus | cgr-miR-1249 | MIMAT0041147 | |
| Dasypus novemcinctus | dno-miR-1249-3p | MIMAT0048032 | |
| View More Rows | |||
| Dog | cfa-miR-1249 | MIMAT0034321 | |
| Eptesicus fuscus | efu-miR-1249 | MIMAT0034894 | |
| Goat | chi-miR-1249 | MIMAT0035921 | |
| Gorilla gorilla | ggo-miR-1249 | MIMAT0024247 | |
| Pan troglodytes | ptr-miR-1249 | MIMAT0007975 | |
| Pig | ssc-miR-1249 | MIMAT0025385 | |
| Pongo pygmaeus | ppy-miR-1249 | MIMAT0016174 | |
| Rabbit | ocu-miR-1249-3p | MIMAT0048586 | |
| Rhesus monkey | mml-miR-1249 | MIMAT0024324 | |
Stem-loop Details
Human
Mouse
Rat
Bovine
Cricetulus griseus
Dasypus novemcinctus
Dog
Eptesicus fuscus
Goat
Gorilla gorilla
Pan troglodytes
Pig
Pongo pygmaeus
Rabbit
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-1249 (v20)
Mouse: mmu-miR-1249 (v17)
Mouse: mmu-miR-1249 (v17)

Genomic Map