Assay Details
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | AAAAGUAUUUGCGGGUUUUGUC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548l | MIMAT0005889 |
| Pan troglodytes | ptr-miR-548l | MIMAT0008226 |
Stem-loop Details
Human
Pan troglodytes

Genomic Map