Assay Details
Gene Family ID: | MIPF0001681, mir-2071 |
Mature miRNA Sequence: | AUUGUUACACCGCGCCGCAAAAG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Branchiostoma floridae | bfl-miR-2071-3p | MIMAT0009999 |
Stem-loop Details
Branchiostoma floridae
More Information
Related Products
miRBase Alias:
Branchiostoma floridae: bfl-miR-2071 (v18)

Genomic Map