Assay Details
Mature miRNA Sequence: | AAAAGCUGGGUUGAGAGGUGA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Xenopus tropicalis | xtr-miR-320 | MIMAT0005564 |
Stem-loop Details
Xenopus tropicalis
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | xtr-miR-320 |
|---|---|
| miRBase Accession Number: | MI0006266 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAAAGCUGGGUUGAGAGGUGA |
| Species: | Xenopus tropicalis |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | AAAAGCUGGGUUGAGAGGUGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Xenopus tropicalis | xtr-miR-320 | MIMAT0005564 |