Assay Details
Mature miRNA Sequence: | AAAAGCUGGGUUGAGAGG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Nomascus leucogenys | nle-miR-320c | MIMAT0049440 |
| Pan troglodytes | ptr-miR-320d | MIMAT0008096 |
Stem-loop Details
Nomascus leucogenys
Pan troglodytes

Genomic Map