Assay Details
Gene Family ID: | MIPF0000174, mir-454 |
Mature miRNA Sequence: | UCCUAACCUUAAGGAUGAG |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Chicken | gga-miR-454-5p | MIMAT0007291 |
Stem-loop Details
Chicken

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | gga-miR-454* |
---|---|
miRBase Accession Number: | MI0006984 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UCCUAACCUUAAGGAUGAG |
Species: | Chicken |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000174, mir-454 |
Mature miRNA Sequence: | UCCUAACCUUAAGGAUGAG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Chicken | gga-miR-454-5p | MIMAT0007291 |