Assay Details
Mature miRNA Sequence: | UUCCUGUCUACUUGAAGCCAC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Physcomitrella patens | ppt-miR1051-3p | MIMAT0005168 |
Stem-loop Details
Physcomitrella patens
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | ppt-miR1051-3p |
|---|---|
| miRBase Accession Number: | MI0006010 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUCCUGUCUACUUGAAGCCAC |
| Species: | Physcomitrella patens |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | UUCCUGUCUACUUGAAGCCAC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Physcomitrella patens | ppt-miR1051-3p | MIMAT0005168 |