Assay Details
Gene Family ID: | MIPF0000254, mir-316 |
Mature miRNA Sequence: | UGUCUUUUUCCGCUUUGCUGCCG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Acyrthosiphon pisum | api-miR-316 | MIMAT0014731 |
| Apis mellifera | ame-miR-316-5p | MIMAT0010122 |
| Polistes canadensis | pca-miR-316-5p | MIMAT0045905 |
Stem-loop Details
Acyrthosiphon pisum
Apis mellifera
Polistes canadensis
More Information
Related Products
miRBase Alias:
Anaerococcus mediterraneensis: ame-miR-316 (v21)

Genomic Map