Assay Details
Gene Family ID: | MIPF0000254, mir-316 |
Mature miRNA Sequence: | UGUCUUUUUCCGCUUACUGACG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila mojavensis | dmo-miR-316 | MIMAT0008627 |
Stem-loop Details
Drosophila mojavensis
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | dmo-miR-316 |
|---|---|
| miRBase Accession Number: | MI0009181 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGUCUUUUUCCGCUUACUGACG |
| Species: | Drosophila mojavensis |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000254, mir-316 |
Mature miRNA Sequence: | UGUCUUUUUCCGCUUACUGACG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila mojavensis | dmo-miR-316 | MIMAT0008627 |