Assay Details
Gene Family ID: | MIPF0000078, mir-187 |
Mature miRNA Sequence: | GCUACAACAGAGGACACAGGA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ornithorhynchus anatinus | oan-miR-187-5p | MIMAT0007266 |
Stem-loop Details
Ornithorhynchus anatinus
More Information
Related Products
miRBase Alias:
Ochrobactrum anthropi: oan-miR-187* (v18)

Genomic Map