Assay Details
Mature miRNA Sequence: | GUACGAUUCGCAUUGCCCAUU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ornithorhynchus anatinus | oan-miR-1330 | MIMAT0006800 |
Stem-loop Details
Ornithorhynchus anatinus
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | oan-miR-1330 |
|---|---|
| miRBase Accession Number: | MI0006664 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GUACGAUUCGCAUUGCCCAUU |
| Species: | Ornithorhynchus anatinus |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | GUACGAUUCGCAUUGCCCAUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ornithorhynchus anatinus | oan-miR-1330 | MIMAT0006800 |