Assay Details
Gene Family ID: | MIPF0000076, mir-190 |
Mature miRNA Sequence: | UGAUAUGUUUGUUAUUUGGUUGGU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Saccoglossus kowalevskii | sko-miR-190 | MIMAT0009634 |
Stem-loop Details
Saccoglossus kowalevskii

Genomic Map