Assay Details
Gene Family ID: | MIPF0000114, mir-375 |
Mature miRNA Sequence: | UUUGUUCGUCCGGCUCGCGUUA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Capitella teleta | cte-miR-375 | MIMAT0009544 |
Stem-loop Details
Capitella teleta
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | cap-miR-375 |
|---|---|
| miRBase Accession Number: | MI0010088 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUUGUUCGUCCGGCUCGCGUUA |
| Species: | Capitella teleta |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000114, mir-375 |
Mature miRNA Sequence: | UUUGUUCGUCCGGCUCGCGUUA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Capitella teleta | cte-miR-375 | MIMAT0009544 |