Assay Details
Mature miRNA Sequence: | ACCCUAUCGAUAUUGUCUCUGCU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Dasypus novemcinctus | dno-miR-454-5p | MIMAT0047904 |
| Ornithorhynchus anatinus | oan-miR-454-5p | MIMAT0007156 |
Stem-loop Details
Dasypus novemcinctus
Ornithorhynchus anatinus
More Information
Related Products
miRBase Alias:
Ochrobactrum anthropi: oan-miR-454* (v18)

Genomic Map