Assay Details
Gene Family ID: | MIPF0000163, mir-320 |
Mature miRNA Sequence: | AAAAGCUGGGUUGAGAGGGU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-320c | MIMAT0005793 |
| Pan troglodytes | ptr-miR-320c | MIMAT0008095 |
| Pongo pygmaeus | ppy-miR-320c | MIMAT0015823 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus

Genomic Map