Assay Details
Gene Family ID: | MIPF0000163, mir-320 |
Mature miRNA Sequence: | AAAAGCUGGGUUGAGAGGA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-320d | MIMAT0006764 |
| Pongo pygmaeus | ppy-miR-320d | MIMAT0015824 |
Stem-loop Details
Human
Pongo pygmaeus

Genomic Map