Assay Details
Mature miRNA Sequence: | UUGAACCCCUGACCUCCU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-2183 | MIMAT0011287 |
Stem-loop Details
Mouse

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | mmu-miR-2183 |
---|---|
miRBase Accession Number: | MI0010833 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UUGAACCCCUGACCUCCU |
Species: | Mouse |
Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | UUGAACCCCUGACCUCCU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-2183 | MIMAT0011287 |