Assay Details
Gene Family ID: | MIPF0000889, mir-2542 |
Mature miRNA Sequence: | UAUUUUUGCUCGGACAUUUAUGU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila pseudoobscura | dps-miR-2542-5p | MIMAT0012340 |
Stem-loop Details
Drosophila pseudoobscura
More Information
Related Products
miRBase Alias:
Derxomyces pseudoschimicola: dps-miR-2542 (v18)

Genomic Map