Assay Details
Gene Family ID: | MIPF0001046, mir-1012 |
Mature miRNA Sequence: | GUGGGUAGAACUUUGAUUAAUA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila simulans | dsi-miR-1012-5p | MIMAT0012425 |
Stem-loop Details
Drosophila simulans
More Information
Related Products
miRBase Alias:
Drosophila simulans: dsi-miR-1012 (v18)

Genomic Map