Assay Details
Gene Family ID: | MIPF0000098, mir-95 |
Mature miRNA Sequence: | UCAACAAACAUUUAUUGUGUGC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Callithrix jacchus | cja-miR-545 | MIMAT0039644 |
| Dasypus novemcinctus | dno-miR-545-3p | MIMAT0047974 |
| Goat | chi-miR-545-3p | MIMAT0036278 |
| Horse | eca-miR-545 | MIMAT0013239 |
Stem-loop Details
Callithrix jacchus
Dasypus novemcinctus
Goat
Horse

Genomic Map