Assay Details
Gene Family ID: | MIPF0000790, mir-2319 |
Mature miRNA Sequence: | UACUCUGUAUUAGGCACUUAG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bovine | bta-miR-2319b | MIMAT0011843 |
Stem-loop Details
Bovine
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | bta-miR-2319b |
|---|---|
| miRBase Accession Number: | MI0011341 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UACUCUGUAUUAGGCACUUAG |
| Species: | Bovine |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000790, mir-2319 |
Mature miRNA Sequence: | UACUCUGUAUUAGGCACUUAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bovine | bta-miR-2319b | MIMAT0011843 |