Assay Details
Gene Family ID: | MIPF0000129, mir-455 |
Mature miRNA Sequence: | UGCAGUCCAUGGGCAUAUACA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Taeniopygia guttata | tgu-miR-455-3p | MIMAT0014610 |
Stem-loop Details
Taeniopygia guttata
More Information
Related Products
miRBase Alias:
Trabulsiella guamensis ATCC 49490: tgu-miR-455 (v18)

Genomic Map