Assay Details
Gene Family ID: | MIPF0001770, mir-2187 |
Mature miRNA Sequence: | UUAAUUAGUAUAGCCUGUUUUA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Zebrafish | dre-miR-2187-5p | MIMAT0011296 |
Stem-loop Details
Zebrafish
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | dre-miR-2187* |
|---|---|
| miRBase Accession Number: | MI0010839 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUAAUUAGUAUAGCCUGUUUUA |
| Species: | Zebrafish |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0001770, mir-2187 |
Mature miRNA Sequence: | UUAAUUAGUAUAGCCUGUUUUA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Zebrafish | dre-miR-2187-5p | MIMAT0011296 |