Assay Details
Gene Family ID: | MIPF0000785, mir-2329 |
Mature miRNA Sequence: | UCUGUGAUGUGAGCUGAUAAGU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Bovine | bta-miR-2329-3p | MIMAT0011859 |
Stem-loop Details
Bovine

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | bta-miR-2329-3p |
---|---|
miRBase Accession Number: | MI0011354 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UCUGUGAUGUGAGCUGAUAAGU |
Species: | Bovine |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000785, mir-2329 |
Mature miRNA Sequence: | UCUGUGAUGUGAGCUGAUAAGU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Bovine | bta-miR-2329-3p | MIMAT0011859 |