Assay Details
Gene Family ID: | MIPF0000242, mir-425 |
Mature miRNA Sequence: | CAUCGGGGAUGUCGUGUCUUU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Taeniopygia guttata | tgu-miR-425-3p | MIMAT0014471 |
Stem-loop Details
Taeniopygia guttata
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | tgu-miR-425-3p |
|---|---|
| miRBase Accession Number: | MI0013657 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAUCGGGGAUGUCGUGUCUUU |
| Species: | Taeniopygia guttata |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000242, mir-425 |
Mature miRNA Sequence: | CAUCGGGGAUGUCGUGUCUUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Taeniopygia guttata | tgu-miR-425-3p | MIMAT0014471 |