Assay Details
Gene Family ID: | MIPF0000838, mir-2240 |
Mature miRNA Sequence: | UCCCAGGGAUUCUCAGUGGAUU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pristionchus pacificus | ppc-miR-2240c | MIMAT0011691 |
Stem-loop Details
Pristionchus pacificus

Genomic Map