Assay Details
Mature miRNA Sequence: | ACAGCACGUGUUACGAUGCUCC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Caenorhabditis elegans | cel-miR-2215-5p | MIMAT0011445 |
Stem-loop Details
Caenorhabditis elegans
More Information
Related Products
miRBase Alias:
Caenorhabditis elegans: cel-miR-2215* (v17)

Genomic Map