Assay Details
Gene Family ID: | MIPF0000787, mir-2209 |
Mature miRNA Sequence: | GAGUGUAACCACUCUUCUCCUUC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Caenorhabditis elegans | cel-miR-2209a-5p | MIMAT0011431 |
Stem-loop Details
Caenorhabditis elegans
More Information
Related Products
miRBase Alias:
Caenorhabditis elegans: cel-miR-2209a* (v17)

Genomic Map