Assay Details
Gene Family ID: | MIPF0000772, mir-2208 |
Mature miRNA Sequence: | AUGCAGAUUUUGGUACACUUCA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Caenorhabditis elegans | cel-miR-2208b-3p | MIMAT0011430 |
Stem-loop Details
Caenorhabditis elegans

Genomic Map