Assay Details
Mature miRNA Sequence: | CAUCGAGACGGUUCCAUACA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bombyx mori | bmo-miR-2761-5p | MIMAT0013634 |
Stem-loop Details
Bombyx mori
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | bmo-miR-2761* |
|---|---|
| miRBase Accession Number: | MI0012315 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAUCGAGACGGUUCCAUACA |
| Species: | Bombyx mori |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | CAUCGAGACGGUUCCAUACA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bombyx mori | bmo-miR-2761-5p | MIMAT0013634 |